Assay Details
| Gene Symbol: KRT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126173.4 | XP_003126221.1 | 1646 | 64 | ||
| GenBank mRNA | AK239722.1 | - | 8 - 9 | 1653 | 64 | |
| AK397301.1 | - | 8 - 9 | 1552 | 64 | ||
| AK397423.1 | - | 3 - 4 | 387 | 64 | ||
| AK400921.1 | - | 7 - 8 | 789 | 64 | ||
More Information
Set Membership: |
|

KRT5
Genomic Map