Assay Details
| Gene Symbol: BASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483806.4 | XP_003483854.1 | 220 | 68 | ||
| GenBank mRNA | AK239723.1 | - | 1 - 2 | 213 | 68 | |
| AK389241.1 | - | 1 - 2 | 213 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003483806.4 transcripts.
Set Membership: |
|

BASP1
Genomic Map