Assay Details
| Gene Symbol: LOC100155195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090310.1 | XP_020945969.1 | 917 | 72 | ||
| GenBank mRNA | AK239735.1 | - | 6 - 7 | 881 | 72 | |
| AK351965.1 | - | 6 - 7 | 869 | 72 | ||
| AK393051.1 | - | 5 - 6 | 744 | 72 | ||
More Information
Set Membership: |
|

LOC100155195
Genomic Map