Assay Details
| Gene Symbol: ZFAND2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927706.4 | XP_001927741.1 | 656 | 65 | ||
| XM_005672224.2 | XP_005672281.1 | 656 | 65 | |||
| GenBank mRNA | AK239736.1 | - | 6 - 7 | 620 | 65 | |
| AK392711.1 | - | 6 - 7 | 656 | 65 | ||
| AY609424.1 | - | 5 - 6 | 636 | 65 | ||
More Information
Set Membership: |
|

ZFAND2B
Genomic Map