Assay Details
| Gene Symbol: PIP4K2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091877.1 | XP_020947536.1 | 445 | 89 | ||
| XM_021091878.1 | XP_020947537.1 | 445 | 89 | |||
| GenBank mRNA | AK239907.1 | - | 1 - 2 | 221 | 89 | |
| AK349843.1 | - | 1 - 2 | 215 | 89 | ||
| AK391581.1 | - | 1 - 2 | 264 | 89 | ||
More Information
Set Membership: |
|

PIP4K2C
Genomic Map