Assay Details
| Gene Symbol: DOP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003480309.4 | XP_003480357.2 | 6981 | 109 | ||
| XM_005659411.3 | XP_005659468.2 | 6952 | 109 | |||
| XM_021091824.1 | XP_020947483.1 | 6981 | 109 | |||
| XM_021091831.1 | XP_020947490.1 | 7579 | 109 | |||
| XM_021091842.1 | XP_020947501.1 | 6952 | 109 | |||
| XM_021091853.1 | XP_020947512.1 | 6971 | 109 | |||
| XM_021091863.1 | XP_020947522.1 | 6981 | 109 | |||
| XM_021091876.1 | XP_020947535.1 | 6971 | 109 | |||
| XM_021091892.1 | XP_020947551.1 | 6971 | 109 | |||
| GenBank mRNA | AK239910.1 | - | 10 - 11 | 1022 | 109 | |
More Information
Set Membership: |
|

DOP1A
Genomic Map