Assay Details
| Gene Symbol: NUTF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126922.5 | XP_003126970.2 | 482 | 91 | ||
| GenBank mRNA | AK239935.1 | - | 3 - 4 | 253 | 91 | |
| AK391986.1 | - | 3 - 4 | 250 | 91 | ||
| AY610424.1 | - | 2 - 3 | 197 | 91 | ||
More Information
Set Membership: |
|

NUTF2
Genomic Map