Assay Details
| Gene Symbol: UTP14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135366.4 | XP_003135414.1 | 2180 | 76 | ||
| XM_005673899.3 | XP_005673956.1 | 2119 | 76 | |||
| GenBank mRNA | AK239956.1 | - | 3 - 4 | 712 | 76 | |
| AK345674.1 | - | 3 - 4 | 647 | 76 | ||
More Information
Set Membership: |
|

UTP14A
Genomic Map