Assay Details
| Gene Symbol: CCN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134452.4 | XP_003134500.1 | 687 | 111 | ||
| XM_003134453.4 | XP_003134501.1 | 664 | 111 | |||
| GenBank mRNA | AK240000.1 | - | 3 - 4 | 400 | 111 | |
More Information
Set Membership: |
|

CCN5
Genomic Map