Assay Details
| Gene Symbol: EFNA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929326.3 | XP_001929361.1 | 207 | 100 | ||
| XM_005663331.3 | XP_005663388.1 | 207 | 100 | |||
| GenBank mRNA | AK240070.1 | - | 1 - 2 | 188 | 100 | |
| AK399409.1 | - | 1 - 2 | 188 | 100 | ||
More Information
Set Membership: |
|

EFNA4
Genomic Map