Assay Details
| Gene Symbol: TRAPPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005657035.3 | XP_005657092.1 | 282 | 86 | ||
| XM_021067851.1 | XP_020923510.1 | 312 | 86 | |||
| GenBank mRNA | AK240117.1 | - | 2 - 3 | 305 | 86 | |
More Information
Set Membership: |
|

TRAPPC1
Genomic Map