Assay Details
| Gene Symbol: PAGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013987719.2 | XP_013843173.1 | 1041 | 127 | ||
| GenBank mRNA | AK240257.1 | - | 2 - 3 | 718 | 127 | |
| AK350237.1 | - | 2 - 3 | 706 | 127 | ||
More Information
Set Membership: |
|

PAGR1
Genomic Map