Assay Details
| Gene Symbol: HSPA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005661654.3 | XP_005661711.1 | 2672 | 86 | ||
| GenBank mRNA | AK240329.1 | - | 2 - 3 | 227 | 86 | |
More Information
Set Membership: |
|

HSPA4
Genomic Map