Assay Details
| Gene Symbol: ISCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003357702.4 | XP_003357750.1 | 209 | 109 | ||
| GenBank mRNA | AK240487.1 | - | 1 - 2 | 160 | 109 | |
More Information
Set Membership: |
|

ISCA1
Genomic Map