Assay Details
| Gene Symbol: MRPS15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095956.1 | XP_020951615.1 | 836 | 84 | ||
| XM_021095957.1 | XP_020951616.1 | 799 | 84 | |||
| XM_021095958.1 | XP_020951617.1 | 755 | 84 | |||
| XM_021095959.1 | XP_020951618.1 | 732 | 84 | |||
| XM_021095960.1 | XP_020951619.1 | 666 | 84 | |||
| GenBank mRNA | AK343256.1 | - | 7 - 8 | 737 | 84 | |
| AY609720.1 | - | 7 - 8 | 714 | 84 | ||
More Information
Set Membership: |
|

MRPS15
Genomic Map