Assay Details
| Gene Symbol: HFE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021098424.1 | XP_020954083.1 | 710 | 62 | ||
| XM_021098425.1 | XP_020954084.1 | 525 | 62 | |||
| XM_021098426.1 | XP_020954085.1 | 685 | 62 | |||
| XM_021098427.1 | XP_020954086.1 | 516 | 62 | |||
| GenBank mRNA | AK343281.1 | - | 2 - 3 | 461 | 62 | |
| AK396311.1 | - | 2 - 3 | 461 | 62 | ||
More Information
Set Membership: |
|

HFE
Genomic Map