Assay Details
| Gene Symbol: ZFAND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021099853.1 | XP_020955512.1 | 2788 | 138 | ||
| XM_021099854.1 | XP_020955513.1 | 2651 | 138 | |||
| XM_021099855.1 | XP_020955514.1 | 2635 | 138 | |||
| GenBank mRNA | AK343390.1 | - | 1 | 1422 | 139 | |
More Information
Set Membership: |
|

ZFAND3
Genomic Map