Assay Details
| Gene Symbol: OTUD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003484107.4 | XP_003484155.2 | 2241 | 68 | ||
| XM_021080668.1 | XP_020936327.1 | 2227 | 68 | |||
| GenBank mRNA | AK343457.1 | - | 2 - 3 | 224 | 68 | |
More Information
Set Membership: |
|

OTUD5
Genomic Map