Assay Details
| Gene Symbol: CPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091972.1 | XP_020947631.1 | 1155 | 104 | ||
| GenBank mRNA | AK343556.1 | - | 2 - 3 | 156 | 104 | |
More Information
Set Membership: |
|

CPM
Genomic Map