Assay Details
| Gene Symbol: CCBE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121722.6 | XP_003121770.3 | 6131 | 82 | ||
| XM_005659840.3 | XP_005659897.1 | 6170 | 82 | |||
| XM_021099891.1 | XP_020955550.1 | 6017 | 82 | |||
| XM_021099895.1 | XP_020955554.1 | 5930 | 82 | |||
| XM_021099901.1 | XP_020955560.1 | 5927 | 82 | |||
| GenBank mRNA | AK343575.1 | - | 1 | 2016 | 82 | |
More Information
Set Membership: |
|

CCBE1
Genomic Map