Assay Details
| Gene Symbol: TRIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127821.5 | XP_003127869.3 | 1143 | 84 | ||
| XM_021095991.1 | XP_020951650.1 | 1143 | 84 | |||
| XM_021095992.1 | XP_020951651.1 | 1143 | 84 | |||
| XM_021095993.1 | XP_020951652.1 | 2548 | 84 | |||
| XM_021095994.1 | XP_020951653.1 | 903 | 84 | |||
| GenBank mRNA | AK343658.1 | - | 9 - 10 | 1131 | 84 | |
More Information
Set Membership: |
|

TRIT1
Genomic Map