Assay Details
| Gene Symbol: LOC100622859 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013979323.2 | XP_013834777.2 | 139 | 72 | ||
| XM_013979324.2 | XP_013834778.2 | 154 | 72 | |||
| GenBank mRNA | AK343932.1 | - | 1 - 2 | 83 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013979323.2, XM_013979324.2 transcripts.
Set Membership: |
|

LOC100622859
Genomic Map