Assay Details
| Gene Symbol: LOC100622859 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013979323.2 | XP_013834777.2 | 1606 | 85 | ||
| XM_021062708.1 | XP_020918367.1 | 2899 | 85 | |||
| XM_021062709.1 | XP_020918368.1 | 2861 | 85 | |||
| GenBank mRNA | AK343932.1 | - | 6 - 7 | 2666 | 85 | |
More Information
Set Membership: |
|

LOC100622859
Genomic Map