Assay Details
Gene Symbol: TMEM254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003133087.4 | XP_003133135.1 | 175 | 62 | ||
GenBank mRNA | AK343944.1 | - | 1 - 2 | 115 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
