Assay Details
| Gene Symbol: RBM48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130141.5 | XP_003130189.3 | 478 | 87 | ||
| GenBank mRNA | AK343984.1 | - | 3 - 4 | 449 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RBM48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130141.5 | XP_003130189.3 | 478 | 87 | ||
| GenBank mRNA | AK343984.1 | - | 3 - 4 | 449 | 87 | |
Set Membership: |
|