Assay Details
| Gene Symbol: WASF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021065503.1 | XP_020921162.1 | 917 | 92 | ||
| XM_021065504.1 | XP_020921163.1 | 830 | 92 | |||
| XM_021065505.1 | XP_020921164.1 | 725 | 92 | |||
| XM_021065506.1 | XP_020921165.1 | 891 | 92 | |||
| XM_021065507.1 | XP_020921166.1 | 625 | 92 | |||
| XM_021065508.1 | XP_020921167.1 | 724 | 92 | |||
| XM_021065509.1 | XP_020921168.1 | 792 | 92 | |||
| XM_021065510.1 | XP_020921169.1 | 625 | 92 | |||
| XM_021065511.1 | XP_020921170.1 | 509 | 92 | |||
| GenBank mRNA | AK343996.1 | - | 5 - 6 | 621 | 92 | |
More Information
Set Membership: |
|

WASF3
Genomic Map