Assay Details
Gene Symbol: SMIM10L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003355574.4 | XP_003355622.1 | 312 | 97 | ||
GenBank mRNA | AK344009.1 | - | 1 | 122 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003355574.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
