Assay Details
| Gene Symbol: CAVIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005652440.3 | XP_005652497.1 | 839 | 71 | ||
| GenBank mRNA | AK344205.1 | - | 1 - 2 | 624 | 71 | |
More Information
Set Membership: |
|

CAVIN2
Genomic Map