Assay Details
| Gene Symbol: CENPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097067.1 | XP_020952726.1 | 1066 | 61 | ||
| XM_021097068.1 | XP_020952727.1 | 1227 | 61 | |||
| GenBank mRNA | AK344429.1 | - | 10 - 11 | 1030 | 61 | |
More Information
Set Membership: |
|

CENPN
Genomic Map