Assay Details
| Gene Symbol: MARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091866.1 | XP_020947525.1 | 1804 | 55 | ||
| XM_021091867.1 | XP_020947526.1 | 1804 | 55 | |||
| GenBank mRNA | AK344458.1 | - | 4 - 5 | 587 | 55 | |
More Information
Set Membership: |
|

MARS1
Genomic Map