Assay Details
| Gene Symbol: B3GNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125112.5 | XP_003125160.2 | 2107 | 89 | ||
| XM_021087479.1 | XP_020943138.1 | 3695 | 89 | |||
| GenBank mRNA | AK344468.1 | - | 1 | 1117 | 89 | |
More Information
Set Membership: |
|

B3GNT2
Genomic Map