Assay Details
| Gene Symbol: LOC100524972 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122663.4 | XP_003122711.2 | 825 | 74 | ||
| XM_003122664.4 | XP_003122712.2 | 696 | 74 | |||
| XM_005660862.3 | XP_005660919.2 | 825 | 74 | |||
| GenBank mRNA | AK344539.1 | - | 5 - 6 | 660 | 74 | |
| AK347083.1 | - | 6 - 7 | 810 | 74 | ||
More Information
Set Membership: |
|

LOC100524972
Genomic Map