Assay Details
| Gene Symbol: PNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929223.4 | XP_001929258.1 | 641 | 70 | ||
| XM_013989064.2 | XP_013844518.2 | 720 | 70 | |||
| XM_021099322.1 | XP_020954981.1 | 729 | 70 | |||
| GenBank mRNA | AK344544.1 | - | 4 - 5 | 583 | 70 | |
| AK397729.1 | - | 4 - 5 | 580 | 70 | ||
More Information
Set Membership: |
|

PNP
Genomic Map