Assay Details
| Gene Symbol: PXYLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013982090.2 | XP_013837544.1 | 416 | 79 | ||
| XM_013982091.2 | XP_013837545.1 | 858 | 79 | |||
| XM_013982093.2 | XP_013837547.1 | 641 | 79 | |||
| XM_013982094.2 | XP_013837548.1 | 232 | 79 | |||
| XM_021069540.1 | XP_020925199.1 | 570 | 79 | |||
| XM_021069541.1 | XP_020925200.1 | 687 | 79 | |||
| GenBank mRNA | AK344565.1 | - | 3 - 4 | 335 | 79 | |
More Information
Set Membership: |
|

PXYLP1
Genomic Map