Assay Details
| Gene Symbol: ARHGAP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021073083.1 | XP_020928742.1 | 2898 | 100 | ||
| GenBank mRNA | AK344611.1 | - | 1 | 1422 | 100 | |
More Information
Set Membership: |
|

ARHGAP22
Genomic Map