Assay Details
| Gene Symbol: NEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481636.4 | XP_003481684.2 | 5854 | 135 | ||
| GenBank mRNA | AK344757.1 | - | 1 | 120 | 135 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: NEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481636.4 | XP_003481684.2 | 5854 | 135 | ||
| GenBank mRNA | AK344757.1 | - | 1 | 120 | 135 | |
Set Membership: |
|