Assay Details
| Gene Symbol: C2H5orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005655017.3 | XP_005655074.1 | 4821 | 91 | ||
| XM_013987605.2 | XP_013843059.1 | 3112 | 91 | |||
| GenBank mRNA | AK344839.1 | - | 1 | 614 | 91 | |
More Information
Set Membership: |
|

C2H5orf24
Genomic Map