Assay Details
| Gene Symbol: LOC100521082 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124818.4 | XP_003124866.2 | 558 | 84 | ||
| GenBank mRNA | AK344849.1 | - | 1 - 2 | 312 | 84 | |
| AK399672.1 | - | 1 - 2 | 317 | 84 | ||
More Information
Set Membership: |
|

LOC100521082
Genomic Map