Assay Details
| Gene Symbol: DNAJA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126994.5 | XP_003127042.1 | 8475 | 77 | ||
| GenBank mRNA | AK344968.1 | - | 4 - 5 | 571 | 77 | |
More Information
Set Membership: |
|

DNAJA2
Genomic Map