Assay Details
| Gene Symbol: GSDMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005653922.3 | XP_005653979.2 | 1046 | 102 | ||
| XM_013979696.2 | XP_013835150.2 | 1012 | 102 | |||
| GenBank mRNA | AK344987.1 | - | 8 - 9 | 970 | 102 | |
| AK345065.1 | - | 8 - 9 | 1004 | 102 | ||
| AK345188.1 | - | 8 - 9 | 1003 | 102 | ||
More Information
Set Membership: |
|

GSDMB
Genomic Map