Assay Details
| Gene Symbol: RPL31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005662362.3 | XP_005662419.1 | 459 | 156 | ||
| GenBank mRNA | AK344989.1 | - | 3 - 4 | 761 | 156 | |
| AK392099.1 | - | 3 - 4 | 752 | 156 | ||
| AK392126.1 | - | 4 - 5 | 394 | 156 | ||
More Information
Set Membership: |
|

RPL31
Genomic Map