Assay Details
| Gene Symbol: C4H8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005655305.3 | XP_005655362.2 | 541 | 112 | ||
| XM_013987984.2 | XP_013843438.1 | 428 | 112 | |||
| XM_013987986.2 | XP_013843440.1 | 428 | 112 | |||
| XM_021090513.1 | XP_020946172.1 | 526 | 112 | |||
| XM_021090514.1 | XP_020946173.1 | 506 | 112 | |||
| XM_021090515.1 | XP_020946174.1 | 428 | 112 | |||
| XM_021090516.1 | XP_020946175.1 | 486 | 112 | |||
| GenBank mRNA | AK345054.1 | - | 2 - 3 | 302 | 112 | |
More Information
Set Membership: |
|

C4H8orf33
Genomic Map