Assay Details
| Gene Symbol: C4H8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005655305.3 | XP_005655362.2 | 625 | 87 | ||
| XM_013987984.2 | XP_013843438.1 | 512 | 87 | |||
| XM_013987986.2 | XP_013843440.1 | 512 | 87 | |||
| XM_021090513.1 | XP_020946172.1 | 610 | 87 | |||
| XM_021090514.1 | XP_020946173.1 | 590 | 87 | |||
| XM_021090515.1 | XP_020946174.1 | 512 | 87 | |||
| XM_021090516.1 | XP_020946175.1 | 570 | 87 | |||
| GenBank mRNA | AK345054.1 | - | 3 - 4 | 386 | 87 | |
More Information
Set Membership: |
|

C4H8orf33
Genomic Map