Assay Details
| Gene Symbol: C4H8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090514.1 | XP_020946173.1 | 729 | 82 | ||
| XM_021090516.1 | XP_020946175.1 | 709 | 82 | |||
| GenBank mRNA | AK345054.1 | - | 4 - 5 | 525 | 82 | |
More Information
Set Membership: |
|

C4H8orf33
Genomic Map