Assay Details
| Gene Symbol: RPS19BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126008.4 | XP_003126056.1 | 379 | 58 | ||
| GenBank mRNA | AK345207.1 | - | 3 - 4 | 286 | 58 | |
| AK346699.1 | - | 3 - 4 | 286 | 58 | ||
More Information
Set Membership: |
|

RPS19BP1
Genomic Map