Assay Details
| Gene Symbol: SUMO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003359010.2 | XP_003359058.1 | 446 | 67 | ||
| GenBank mRNA | AK345302.1 | - | 3 - 4 | 334 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SUMO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003359010.2 | XP_003359058.1 | 446 | 67 | ||
| GenBank mRNA | AK345302.1 | - | 3 - 4 | 334 | 67 | |
Set Membership: |
|