Assay Details
| Gene Symbol: PGP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481109.4 | XP_003481157.2 | 724 | 62 | ||
| GenBank mRNA | AK345409.1 | - | 1 - 2 | 714 | 62 | |
| AY610024.1 | - | 1 - 2 | 174 | 62 | ||
More Information
Set Membership: |
|

PGP
Genomic Map