Assay Details
| Gene Symbol: BAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925236.6 | XP_001925271.3 | 2732 | 92 | ||
| XM_005669632.3 | XP_005669689.1 | 2628 | 92 | |||
| GenBank mRNA | AK345438.1 | - | 1 | 123 | 92 | |
More Information
Set Membership: |
|

BAP1
Genomic Map