Assay Details
| Gene Symbol: ZNF577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013995217.2 | XP_013850671.1 | 506 | 57 | ||
| GenBank mRNA | AK345787.1 | - | 3 - 4 | 400 | 57 | |
More Information
Set Membership: |
|

ZNF577
Genomic Map