Assay Details
| Gene Symbol: ZFAND2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124245.4 | XP_003124293.1 | 343 | 66 | ||
| XM_005655063.3 | XP_005655120.1 | 430 | 66 | |||
| XM_005655064.3 | XP_005655121.1 | 252 | 66 | |||
| GenBank mRNA | AK345938.1 | - | 2 - 3 | 281 | 66 | |
| AK348541.1 | - | 2 - 3 | 284 | 66 | ||
More Information
Set Membership: |
|

ZFAND2A
Genomic Map